missing translation for 'onlineSavingsMsg'
Learn More

Denatonium Benzoate, Anhydrous, Granular, NF, 99.5-101%, Spectrum™ Chemical

Catalog No. p-7157338
Change view
Click to view available options
Quantity:
5 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602040 5 g Amber Glass Bottle
18602042 1 kg Amber Glass Bottle
18602043 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602040 Supplier Spectrum Chemical Mfg Cor Supplier No. DE1015GM
Only null left

DE101, 3734-33-6, C28H34N2O3

Spectrum™ Chemical Denatonium Benzoate, Anhydrous, Granular, NF is used as a denaturant and masking agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C28H34N2O3
Molecular Weight (g/mol) 446.59
MDL Number MFCD00031578
InChI Key VWTINHYPRWEBQY-UHFFFAOYSA-N
IUPAC Name benzyl({[(2,6-dimethylphenyl)carbamoyl]methyl})diethylazanium benzoate
SMILES [O-]C(=O)C1=CC=CC=C1.CC[N+](CC)(CC(=O)NC1=C(C)C=CC=C1C)CC1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Melting Range 163-170°C
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 99.5 to 101%
Grade NF