missing translation for 'onlineSavingsMsg'
Learn More

Demethoxycurcumin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018239
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018239 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018239 Supplier Merck Emd Millipore Supplier No. 9059310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Food and Beverages

Specifications

CAS 22608-11-3
Physical Form Neat
Linear Formula C20H18O5
MDL Number MFCD03427310
Synonym (E,E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione; Curcumin II; Desmethoxycurcumin; Monodemethoxycurcumin
Type Analytical Standard
Quantity 10 mg
Formula Weight 338.35
Percent Purity ≥95% (HPLC)
Grade Analytical Standard