Learn More
Demeclocycline hydrochloride
CAS: 64-73-3 | C21H22Cl2N2O8 | 501.313 g/mol
Supplier: Thermo Scientific Chemicals J63102MD
Description
Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDemeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
| Demeclocycline hydrochloride | |
| >245°C (decomposition) | |
| C21H22Cl2N2O8 | |
| 250 mg | |
| 14,2886 | |
| Soluble in water. | |
| CN(C)C1C2CC3C(C4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)Cl)O.Cl | |
| 501.313 | |
| 501.32 |
| 64-73-3 | |
| Yellow | |
| MFCD00082371 | |
| 3849198 | |
| demeclocycline hydrochloride, demeclocycline hcl, declomycin, detravis, ledermycine, clortetrin, ledermycin, ledermicina, veraciclina, demeplus | |
| OAPVUSSHCBRCOL-KBHRXELFSA-N | |
| (4S,4aS,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride | |
| 54686764 | |
| Powder |
Chemical Identifiers
| 64-73-3 | |
| 501.313 | |
| OAPVUSSHCBRCOL-KBHRXELFSA-N | |
| 54686764 | |
| CN(C)C1C2CC3C(C4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)Cl)O.Cl |
| C21H22Cl2N2O8 | |
| MFCD00082371 | |
| demeclocycline hydrochloride, demeclocycline hcl, declomycin, detravis, ledermycine, clortetrin, ledermycin, ledermicina, veraciclina, demeplus | |
| (4S,4aS,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 200-592-2
RTECSNumber : QI7700000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only