accessibility menu, dialog, popup

UserName

CAS RN 64-73-3

IUPAC Name: hydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Synonyms: demeclocycline
Molecular Weight (g/mol): 501.31
Molecular Formula: C21H22Cl2N2O8
InChi Key: POPIIQNWBBGGRT-YHBXTMLJSA-N
SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3[C@H](O)C4=C(Cl)C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results