missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Deluxe Office Monitor Arm

Deluxe monitor arm effortlessly adjusts to the angle and focal depth of your flat screen monitor for either seated or standing applications.

Supplier:  Fisherbrand™ FS203S

 View more versions of this product

Catalog No. 06001039


Only null left
Explore available promotions

Description

Description

  • 4 to 21 in. horizontal travel range
  • Accommodates all VESA-compliant monitors weighting up to 25 lb.
  • In clamp or grommet mounts
  • Portrait or landscape monitor compatible
Specifications

Specifications

4 in.
10.2 cm
Double Arm
45.7 cm
18 in.
Silver
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions