missing translation for 'onlineSavingsMsg'
Learn More

Deltamethrin 97.0+%, TCI America™

Catalog No. D4775100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4775100MG 100 mg
D47751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4775100MG Supplier TCI America Supplier No. D4775100MG

Chemical Identifiers

CAS 52918-63-5
Molecular Formula C22H19Br2NO3
Molecular Weight (g/mol) 505.206
MDL Number MFCD00870122
InChI Key OWZREIFADZCYQD-FLXSOZOKSA-N
Synonym deltamethrin
PubChem CID 13013599
IUPAC Name [(S)-cyano-(3-phenoxyphenyl)methyl] (1R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C(C1C(=O)OC(C#N)C2=CC(=CC=C2)OC3=CC=CC=C3)C=C(Br)Br)C

Specifications

Melting Point 101°C
Quantity 100 mg
UN Number 2811
Formula Weight 505.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Deltamethrin
RTECSNumber GZ1233000
TSCA No
Recommended Storage Freezer