accessibility menu, dialog, popup

UserName

CAS RN 52918-63-5

IUPAC Name: (S)-cyano(3-phenoxyphenyl)methyl (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Synonyms: stricker decis 0.5ulv suspend crackdown zorcis DeltaGard decis deltagran deltametryna decamethrin
Molecular Weight (g/mol): 505.21
Molecular Formula: C22H19Br2NO3
InChi Key: OWZREIFADZCYQD-NSHGMRRFSA-N
SMILES: CC1(C)[C@@H](C=C(Br)Br)[C@H]1C(=O)O[C@H](C#N)C1=CC=CC(OC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results