missing translation for 'onlineSavingsMsg'
Learn More

delta-Valerolactone 98.0+%, TCI America™

Supplier:  TCI America V003925G

 View more versions of this product

Catalog No. V003925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

delta-Valerolactone
Colorless
C5H8O2
25 g
OZJPLYNZGCXSJM-UHFFFAOYSA-N
oxan-2-one
10953
100.12
Liquid
542-28-9
220°C
MFCD00006645
delta-valerolactone, tetrahydro-2h-pyran-2-one, 5-valerolactone, 2h-pyran-2-one, tetrahydro, tetrahydropyran-2-one, tetrahydro-2-pyranone, penta-1,5-lactone, .delta.-valerolactone, .delta.-valeryllactone, tetrahydro-pyran-2-one
O=C1CCCCO1
100.12
CHEBI:16545
≥98.0% (GC)

Chemical Identifiers

542-28-9
100.12
OZJPLYNZGCXSJM-UHFFFAOYSA-N
10953
oxan-2-one
C5H8O2
MFCD00006645
delta-valerolactone, tetrahydro-2h-pyran-2-one, 5-valerolactone, 2h-pyran-2-one, tetrahydro, tetrahydropyran-2-one, tetrahydro-2-pyranone, penta-1,5-lactone, .delta.-valerolactone, .delta.-valeryllactone, tetrahydro-pyran-2-one
CHEBI:16545
O=C1CCCCO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5536

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions