accessibility menu, dialog, popup

UserName

OO

CAS RN 542-28-9

IUPAC Name: oxan-2-one
Synonyms: delta-valerolactone|5-valerolactone|penta-1,5-lactone|delta-valeryllactone
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: OZJPLYNZGCXSJM-UHFFFAOYSA-N
SMILES: O=C1CCCCO1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)