missing translation for 'onlineSavingsMsg'
Learn More

delta-Valerolactone 98.0+%, TCI America™

Catalog No. V0039500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0039500G 500 g
V003925G 25 g
V0039100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V0039500G Supplier TCI America Supplier No. V0039500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 542-28-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00006645
InChI Key OZJPLYNZGCXSJM-UHFFFAOYSA-N
Synonym delta-valerolactone, tetrahydro-2h-pyran-2-one, 5-valerolactone, 2h-pyran-2-one, tetrahydro, tetrahydropyran-2-one, tetrahydro-2-pyranone, penta-1,5-lactone, .delta.-valerolactone, .delta.-valeryllactone, tetrahydro-pyran-2-one
PubChem CID 10953
ChEBI CHEBI:16545
IUPAC Name oxan-2-one
SMILES O=C1CCCCO1

Specifications

Color Colorless
Boiling Point 220°C
Quantity 500 g
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material delta-Valerolactone
EINECSNumber (5)-5536
TSCA Yes
Recommended Storage Freezer