missing translation for 'onlineSavingsMsg'
Learn More

delta-Valerolactone, 98%, may contain polymer

Catalog No. AAL0479414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0479414 25 g
AAL0479422 100 g
AAL0479436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0479414 Supplier Thermo Scientific Chemicals Supplier No. L0479414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 542-28-9 | C5H8O2 | 100.12 g/mol

δ-Valerolactone is employed as a chemical intermediate in the synthesis of polymers. It is also used to synthesize the copolyesters by lipase-catalyzed ring opening polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
δ-Valerolactone is employed as a chemical intermediate in the synthesis of polymers. It is also used to synthesize the copolyesters by lipase-catalyzed ring opening polymerization.

Solubility
Miscible with water.

Notes
Store in a tightly closed container. Avoid breathing dust, vapor, mist, or gas.

Chemical Identifiers

CAS 542-28-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00006645
InChI Key OZJPLYNZGCXSJM-UHFFFAOYSA-N
Synonym delta-valerolactone, tetrahydro-2h-pyran-2-one, 5-valerolactone, 2h-pyran-2-one, tetrahydro, tetrahydropyran-2-one, tetrahydro-2-pyranone, penta-1,5-lactone, .delta.-valerolactone, .delta.-valeryllactone, tetrahydro-pyran-2-one
PubChem CID 10953
ChEBI CHEBI:16545
IUPAC Name oxan-2-one
SMILES O=C1CCCCO1

Specifications

Melting Point -13°C to -12°C
Density 1.056
Boiling Point 58°C to 60°C (0.5 mmHg)
Flash Point 112°C (233°F)
Refractive Index 1.458
Quantity 25 g
Beilstein 106436
Solubility Information Miscible with water.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material delta-Valerolactone, may contain polymer; distil to remove
Show More Show Less
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 208-807-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only