Learn More
delta-Octanolactone, 97%
CAS: 698-76-0 | C8H14O2 | 142.198 g/mol
Supplier: Thermo Scientific Chemicals L1430322
Description
delta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsdelta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
| delta-Octanolactone | |
| -14°C | |
| 238°C | |
| C8H14O2 | |
| MFCD00144051 | |
| 111515 | |
| Not miscible or difficult to mix in water. | |
| CCCC1CCCC(=O)O1 | |
| 142.198 | |
| 142.2 |
| 698-76-0 | |
| 1.002 | |
| 125°C (257°F) | |
| 1.455 | |
| 100 g | |
| delta-octanolactone, delta-octalactone, 5-octanolide, 5-hydroxyoctanoic acid lactone, 2h-pyran-2-one, tetrahydro-6-propyl, 5-octalactone, .delta.-octalactone, tetrahydro-6-propyl-2h-pyran-2-one, delta-octanolide, fema no. 3214 | |
| FYTRVXSHONWYNE-UHFFFAOYSA-N | |
| 6-propyloxan-2-one | |
| 12777 | |
| 97% |
Chemical Identifiers
| 698-76-0 | |
| 142.198 | |
| FYTRVXSHONWYNE-UHFFFAOYSA-N | |
| 12777 | |
| CCCC1CCCC(=O)O1 |
| C8H14O2 | |
| MFCD00144051 | |
| delta-octanolactone, delta-octalactone, 5-octanolide, 5-hydroxyoctanoic acid lactone, 2h-pyran-2-one, tetrahydro-6-propyl, 5-octalactone, .delta.-octalactone, tetrahydro-6-propyl-2h-pyran-2-one, delta-octanolide, fema no. 3214 | |
| 6-propyloxan-2-one |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 211-820-5
RTECSNumber : UQ1355500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only