accessibility menu, dialog, popup

UserName

CAS RN 698-76-0

IUPAC Name: 6-propyloxan-2-one
Synonyms: delta-octalactone
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: FYTRVXSHONWYNE-UHFFFAOYNA-N
SMILES: CCCC1CCCC(=O)O1
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)