missing translation for 'onlineSavingsMsg'
Learn More

delta-Octanolactone, 97%

Catalog No. AAL1430314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1430314 25 g
AAL1430322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1430314 Supplier Thermo Scientific Chemicals Supplier No. L1430314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 698-76-0 | C8H14O2 | 142.198 g/mol

delta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 698-76-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00144051
InChI Key FYTRVXSHONWYNE-UHFFFAOYSA-N
Synonym delta-octanolactone, delta-octalactone, 5-octanolide, 5-hydroxyoctanoic acid lactone, 2h-pyran-2-one, tetrahydro-6-propyl, 5-octalactone, .delta.-octalactone, tetrahydro-6-propyl-2h-pyran-2-one, delta-octanolide, fema no. 3214
PubChem CID 12777
IUPAC Name 6-propyloxan-2-one
SMILES CCCC1CCCC(=O)O1

Specifications

Melting Point -14°C
Density 1.002
Boiling Point 238°C
Flash Point 125°C (257°F)
Refractive Index 1.455
Quantity 25 g
Beilstein 111515
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material delta-Octanolactone
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 211-820-5
RTECSNumber UQ1355500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only