missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ delta Catenin Monoclonal Antibody (K29002_1E2), Invitrogen™

Catalog No. MA556187
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556187 200 μL
MA555807 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556187 Supplier Invitrogen™ Supplier No. MA556187
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: YIRSALRQEK ALSAIADLLT NEHERVVKAA SGALRNLAVD ARNKELIGKH AIPNLVKNLP GGQQNSSWNF SEDTVISILN TINEVIAENL EAAKKLRETQ GIEKLVLINK SGNRSEKEVR AAALVLQTIW GYKELRKPLE KEGWKKSDFQ VNLNNASRSQ SSHSYDDSTL PLIDRNQKSD KKPDREEIQM SNMGSNTKSL DNNYSTPNER GDHNRTLDRS GDLGDMEPLK GTTPLMQDEG

Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.

Specifications

Antigen delta Catenin
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K29002_1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CTNND1
Gene Accession No. O60716
Gene Alias AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN
Gene Symbols CTNND1
Host Species Mouse
Immunogen Recombinant human CTNND1 (immunogen range is 701-940).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1500
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less