missing translation for 'onlineSavingsMsg'
Learn More

Deionized Water, MDA, APHA, EPA, AOAC Reagent Grade, Reagents

Catalog No. p-200020998
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336888 10 L
01336887 200 L
01336886 20 L
01336885 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336888 Supplier Labchem Inc Supplier No. C234640010F
Only null left

Deionized Water, MDA, APHA, EPA, AOAC Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Oxide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Percent Purity 100%
Packaging Cubitainer
Sterility Non-sterile
Quantity 10 L
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Deionized Water, MDA, APHA, EPA, AOAC Reagent Grade