missing translation for 'onlineSavingsMsg'
Learn More

Deionized Water, MDA, APHA, EPA, AOAC Reagent Grade, Reagents

Catalog No. 01336888
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336888 10 L
01336887 200 L
01336886 20 L
01336885 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336888 Supplier Labchem Inc Supplier No. C234640010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Deionized Water, MDA, APHA, EPA, AOAC Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Oxide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Percent Purity 100%
Packaging Cubitainer
Sterility Non-sterile
Quantity 10 L
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Deionized Water, MDA, APHA, EPA, AOAC Reagent Grade