missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydrocostus lactone, ≥98.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 477-43-0 |
| Linear Formula | C15H18O2 |
| MDL Number | MFCD00210277 |
| Synonym | (-)-Dehydrocostus lactone; Dehydrocostus lactone; Epiligulyl oxide; (3 AS,6 aR,9 aR,9 bS)-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3 H)-one; Dehydro-costus lactone (6 CI,7 CI); Dehydrocostuslactone; Guaia-4(15),10(14),11(13)-trien-12-oic acid, 6 alpha-hydroxy-, gamma-lactone; [3 AS-(3 alpha,6 aalpha,9 aalpha,9 bbeta)]-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3 H)-one |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 230.30 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Analytical Standard |