Learn More
Dehydroacetic acid, 98%
CAS: 520-45-6 | C8H8O4 | 168.148 g/mol
Supplier: Thermo Scientific Chemicals A178270B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dehydroacetic acid | |
| 109°C to 113°C | |
| 157°C (314°F) | |
| MFCD00066709 | |
| 6129 | |
| dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4 | |
| CC1=CC(=O)C(C(=O)O1)C(=O)C | |
| 168.148 | |
| 168.15 |
| 520-45-6 | |
| 269°C to 270°C | |
| C8H8O4 | |
| 1000 g | |
| 14,2865 | |
| PGRHXDWITVMQBC-UHFFFAOYSA-N | |
| 3-acetyl-6-methylpyran-2,4-dione | |
| 122903 | |
| 98% |
Chemical Identifiers
| 520-45-6 | |
| 168.148 | |
| PGRHXDWITVMQBC-UHFFFAOYSA-N | |
| 122903 | |
| CC1=CC(=O)C(C(=O)O1)C(=O)C |
| C8H8O4 | |
| MFCD00066709 | |
| dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4 | |
| 3-acetyl-6-methylpyran-2,4-dione |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 208-293-9
RTECSNumber : UP8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only