missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dehydroabietylamine 90.0+%, TCI America™

Catalog No. D15885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15885G 5 g
D158825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15885G Supplier TCI America Supplier No. D15885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1446-61-3
Molecular Formula C20H31N
Molecular Weight (g/mol) 285.475
MDL Number MFCD00213430
InChI Key JVVXZOOGOGPDRZ-SLFFLAALSA-N
Synonym dehydroabietylamine, +-dehydroabietylamine, 1r,4as,10ar-7-isopropyl-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl methanamine, amine d, dehydroabiethylamine, leelamine, podocarpa-8,11,13-trien-15-amine, 13-isopropyl, 1-phenanthrenemethanamine, 1,2,3,4,4a,9,10,10a-octahydro-1,4a-dimethyl-7-1-methylethyl-, 1r,4as,10ar, pubchem7951, 4-12-00-03005 beilstein handbook reference
PubChem CID 62034
IUPAC Name [(1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthren-1-yl]methanamine
SMILES CC(C)C1=CC2=C(C=C1)C3(CCCC(C3CC2)(C)CN)C

Specifications

Melting Point 40°C
Quantity 5 g
Formula Weight 285.48
Percent Purity ≥90.0% (GC)
Chemical Name or Material (+)-Dehydroabietylamine [Optical Resolving Agent]
EINECSNumber (4)-1242&(7)-0939
TSCA Yes