missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dehydroabietylamine hydrochloride

Catalog No. AAJ63123MA
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63123MA 10 mg
AAJ63123MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63123MA Supplier Thermo Scientific Chemicals Supplier No. J63123MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16496-99-4 | C20H32ClN | 321.93 g/mol

Resolving agent for carboxylic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16496-99-4
Molecular Formula C20H32ClN
Molecular Weight (g/mol) 321.93
MDL Number MFCD06795849
InChI Key CVPQLGCAWUAYPF-UHFFFAOYNA-N
Synonym leelamine hydrochloride, dehydroabiethylamine hydrochloride, +-dehydroabietylamine, lylamine hydrochloride, nsc 2955 hydrochloride, +-dehydroabietylamine hydrochloride, +-dehydroabiethylamine hydrochloride, abieta-8,11,13-trien-18-amine-hydrogen chloride 1/1, 13-isopropyl-podocarpa-8,11,13-trien-15-amine hydrochloride, 1r,4as,10ar-7-isopropyl-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl methanamine hydrochloride
PubChem CID 16759156
IUPAC Name [(1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthren-1-yl]methanamine;hydrochloride
SMILES Cl.CC(C)C1=CC2=C(C=C1)C1(C)CCCC(C)(CN)C1CC2

Specifications

Packaging Plastic bottle
Quantity 10 mg
Beilstein 3084620
Formula Weight 321.9
Physical Form Solid
Chemical Name or Material (+)-Dehydroabietylamine hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only