missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Deferoxamine Mesylate, Calbiochem™,

Catalog No. 2527501gm
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2527501GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2527501GM Supplier MilliporeSigma™ Supplier No. 2527501GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Parenteral iron chelating agent

Chemical Identifiers

CAS 138-14-7
Molecular Formula C26H52N6O11S
Molecular Weight (g/mol) 656.793
InChI Key IDDIJAWJANBQLJ-UHFFFAOYSA-N
Synonym deferoxamine mesylate, desferal, deferoxamine mesilate, desferrioxamine b mesylate, desferrioxamine mesylate, desferal mesylate, deferoxamine b mesylate, deferoxamine methanesulfonate, desferrioxamine methanesulfonate
PubChem CID 62881
ChEBI CHEBI:31460
IUPAC Name N-[5-[[4-[5-[acetyl(hydroxy)amino]pentylamino]-4-oxobutanoyl]-hydroxyamino]pentyl]-N'-(5-aminopentyl)-N'-hydroxybutanediamide;methanesulfonic acid
SMILES CC(=O)N(CCCCCNC(=O)CCC(=O)N(CCCCCNC(=O)CCC(=O)N(CCCCCN)O)O)O.CS(=O)(=O)O

Specifications

Melting Point 148°C to 149°C
Color Yellow
Quantity 1 g
Solubility Information Partially soluble in cold water
Formula Weight 656.8
Physical Form Solid
Chemical Name or Material Deferoxamine Mesylate
Recommended Storage 2° to 8°C (35.6° to 46.4°F)