missing translation for 'onlineSavingsMsg'
Learn More

Deferasirox System Suitabiltiy Mixture, MilliporeSigma™ Supelco™

Catalog No. 111002490
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002490 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002490 Supplier Merck Emd Millipore Supplier No. 116566720MG
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 201530-41-8
Type USP Reference Standard
Quantity 20 mg
Grade Pharmaceutical Primary Standard