missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Defensin 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149952
Description
Defensin 5 Polyclonal Antibody for IHC, Western Blot
Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several of the alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 5, is highly expressed in the secretory granules of Paneth cells of the ileum.Chemical Identifiers
| Defensin 5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q01523 | |
| 1670 | |
| Recombinant Defensin Alpha 5, Paneth Cell Specific (DEFa5) (Glu20-Arg94). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DEFA5 | |
| DEF5; DEFA5; defensin 5; defensin alpha 5; Defensin, alpha 5; defensin, alpha 5, Paneth cell-specific; defensin, alpha 5, preproprotein; defensin-5; HD5; HD-5; HD5(20-94); HD5(23-94); HD5(29-94); HD5(56-94); HD5(63-94); MGC129728 | |
| DEFA5 | |
| Primary | |
| Protein A |
Specifications
| Defensin 5 | |
| Polyclonal | |
| Unconjugated | |
| DEFA5 | |
| DEF5; DEFA5; defensin 5; defensin alpha 5; Defensin, alpha 5; defensin, alpha 5, Paneth cell-specific; defensin, alpha 5, preproprotein; defensin-5; HD5; HD-5; HD5(20-94); HD5(23-94); HD5(29-94); HD5(56-94); HD5(63-94); MGC129728 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1670 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q01523 | |
| DEFA5 | |
| Recombinant Defensin Alpha 5, Paneth Cell Specific (DEFa5) (Glu20-Arg94). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |