missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethylammonium Bromide 99.0+%, TCI America™

Supplier:  TCI America D1467500G

 View more versions of this product

Catalog No. D1467500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Decyltrimethylammonium Bromide
242°C
4.0 to 7.5 (50g/L H2O 25°C)
MFCD00041973
decyltrimethylammonium bromide, n,n,n-trimethyldecan-1-aminium bromide, n-decyltrimethylammonium bromide, trimethyldecylammonium bromide, n,n,n-trimethyldecylammonium bromide, n,n,n-trimethyl-1-decanaminium bromide, 1-decanaminium, n,n,n-trimethyl-, bromide, decyl trimethyl azanium bromide, fsm 20, decyl trimethyl ammonium bromide
[Br-].CCCCCCCCCC[N+](C)(C)C
280.29
CHEBI:295756
≥99.0% (T)
2082-84-0
White
C13H30BrN
500 g
PLMFYJJFUUUCRZ-UHFFFAOYSA-M
decyltrimethylazanium bromide
16388
280.29
Crystalline Powder

Chemical Identifiers

2082-84-0
280.29
PLMFYJJFUUUCRZ-UHFFFAOYSA-M
16388
decyltrimethylazanium bromide
C13H30BrN
MFCD00041973
decyltrimethylammonium bromide, n,n,n-trimethyldecan-1-aminium bromide, n-decyltrimethylammonium bromide, trimethyldecylammonium bromide, n,n,n-trimethyldecylammonium bromide, n,n,n-trimethyl-1-decanaminium bromide, 1-decanaminium, n,n,n-trimethyl-, bromide, decyl trimethyl azanium bromide, fsm 20, decyl trimethyl ammonium bromide
CHEBI:295756
[Br-].CCCCCCCCCC[N+](C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0184&(9)-1971

RTECSNumber : BP6352000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions