accessibility menu, dialog, popup

UserName

CAS RN 2082-84-0

IUPAC Name: decyltrimethylazanium bromide
Synonyms: decyltrimethylammonium bromide
Molecular Weight (g/mol): 280.29
Molecular Formula: C13H30BrN
InChi Key: PLMFYJJFUUUCRZ-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCC[N+](C)(C)C
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)