missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethylammonium Bromide 99.0+%, TCI America™

Catalog No. D146725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D146725G 25 g
D1467500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D146725G Supplier TCI America Supplier No. D146725G

Chemical Identifiers

CAS 2082-84-0
Molecular Formula C13H30BrN
Molecular Weight (g/mol) 280.29
MDL Number MFCD00041973
InChI Key PLMFYJJFUUUCRZ-UHFFFAOYSA-M
Synonym decyltrimethylammonium bromide, n,n,n-trimethyldecan-1-aminium bromide, n-decyltrimethylammonium bromide, trimethyldecylammonium bromide, n,n,n-trimethyldecylammonium bromide, n,n,n-trimethyl-1-decanaminium bromide, 1-decanaminium, n,n,n-trimethyl-, bromide, decyl trimethyl azanium bromide, fsm 20, decyl trimethyl ammonium bromide
PubChem CID 16388
ChEBI CHEBI:295756
IUPAC Name decyltrimethylazanium bromide
SMILES [Br-].CCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 242°C
Color White
pH 4.0 to 7.5 (50g/L H2O 25°C)
Quantity 25 g
Formula Weight 280.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Decyltrimethylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
RTECSNumber BP6352000
TSCA Yes