missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethylammonium bromide, 99%

Catalog No. AC406230250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406230250 25 g Glass Bottle
AC406231000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406230250 Supplier Thermo Scientific Chemicals Supplier No. 406230250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2082-84-0 | C13H30BrN | 280.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 2082-84-0
Molecular Formula C13H30BrN
Molecular Weight (g/mol) 280.29
MDL Number MFCD00041973
InChI Key PLMFYJJFUUUCRZ-UHFFFAOYSA-M
Synonym decyltrimethylammonium bromide, n,n,n-trimethyldecan-1-aminium bromide, n-decyltrimethylammonium bromide, trimethyldecylammonium bromide, n,n,n-trimethyldecylammonium bromide, n,n,n-trimethyl-1-decanaminium bromide, 1-decanaminium, n,n,n-trimethyl-, bromide, decyl trimethyl azanium bromide, fsm 20, decyl trimethyl ammonium bromide
PubChem CID 16388
ChEBI CHEBI:295756
IUPAC Name decyl(trimethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCC[N+](C)(C)C

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3(CH2)9N(CH3)3Br
Quantity 25 g
Formula Weight 280.28
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material Decyltrimethylammonium bromide, 99%
Hazard Category PRODUCT NEEDS INERT ATMOSPHERE
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-219-7
RTECSNumber BP6352000
TSCA TSCA

RUO – Research Use Only