missing translation for 'onlineSavingsMsg'
Learn More

Decylphosphonic Acid 98.0+%, TCI America™

Catalog No. p-7127971
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38841G 1 g
D38845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38841G Supplier TCI America Supplier No. D38841G
Only null left

Chemical Identifiers

CAS 6874-60-8
Molecular Formula C10H23O3P
Molecular Weight (g/mol) 222.265
MDL Number MFCD00015830
InChI Key DZQISOJKASMITI-UHFFFAOYSA-N
Synonym decanephosphonic acid, phosphonic acid, decyl, 1-decanephosphonic acid, decyl-phosphonic acid, 1-decylphosphonic acid, n-decylphosphonic acid, phosphonic acid, p-decyl, decylphosphonicacid, n-decanephosphonic acid, acmc-1cuhv
PubChem CID 81309
IUPAC Name decylphosphonic acid
SMILES CCCCCCCCCCP(=O)(O)O

Specifications

Melting Point 102°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 222.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Decylphosphonic Acid
RTECSNumber SZ7525000
TSCA Yes