accessibility menu, dialog, popup

UserName

H3CPOHOHO

CAS RN 6874-60-8

IUPAC Name: decylphosphonic acid
Synonyms: phosphonic acid, decyl-
Molecular Weight (g/mol): 222.27
Molecular Formula: C10H23O3P
InChi Key: DZQISOJKASMITI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCP(O)(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)