missing translation for 'onlineSavingsMsg'
Learn More

Decyldimethylchlorosilane, 97%, Thermo Scientific™

Catalog No. AC432190050
Change view
Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432190050 5mL Glass bottle
AC432190250 25mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432190050 Supplier Thermo Scientific Chemicals Supplier No. 432190050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38051-57-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.88
InChI Key ZLZGHBNDPINFKG-UHFFFAOYSA-N
Synonym chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa
PubChem CID 2758020
IUPAC Name chloro-decyl-dimethylsilane
SMILES CCCCCCCCCC[Si](C)(C)Cl

Specifications

Density 0.8660g/mL
Boiling Point 98°C (3.0 hPa)
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Quantity 5mL
Specific Gravity 0.866
Formula Weight 234.88
Percent Purity 97%
Chemical Name or Material Decyldimethylchlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 253-761-8

RUO – Research Use Only