missing translation for 'onlineSavingsMsg'
Learn More

Decyl Acetate 98.0+%, TCI America™

Catalog No. A136625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A136625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A136625ML Supplier TCI America Supplier No. A136625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-17-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00026538
InChI Key NUPSHWCALHZGOV-UHFFFAOYSA-N
Synonym n-decyl acetate, acetic acid, decyl ester, n-decyl ethanoate, 1-decanol acetate, decyl ethanoate, acetate c 10, decanyl acetate, 1-decyl acetate, acetic acid n-decyl ester, decanol acetate
PubChem CID 8167
IUPAC Name decyl acetate
SMILES CCCCCCCCCCOC(C)=O

Specifications

Color Colorless
Boiling Point 248°C
Quantity 25 mL
Formula Weight 200.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decyl Acetate
EINECSNumber (2)-0735
RTECSNumber AG5235000
TSCA Yes