missing translation for 'onlineSavingsMsg'
Learn More

Decoquinate 97.0+%, TCI America™

Catalog No. D41921G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41921G Supplier TCI America Supplier No. D41921G
Only null left

Chemical Identifiers

CAS 18507-89-6
Molecular Formula C24H35NO5
Molecular Weight (g/mol) 417.546
MDL Number MFCD00673686
InChI Key JHAYEQICABJSTP-UHFFFAOYSA-N
Synonym 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester, Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate
PubChem CID 29112
IUPAC Name ethyl 6-decoxy-7-ethoxy-4-oxo-1H-quinoline-3-carboxylate
SMILES CCCCCCCCCCOC1=C(C=C2C(=C1)C(=O)C(=CN2)C(=O)OCC)OCC

Specifications

Melting Point 243°C
Color Yellow
Quantity 1 g
Formula Weight 417.55
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Decoquinate
RTECSNumber VB1983800
TSCA No