accessibility menu, dialog, popup

UserName

H3COONHOH3COOCH3

CAS RN 18507-89-6

IUPAC Name: ethyl 6-(decyloxy)-7-ethoxy-4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: decoquinate decoquinatum deccox (product) decoquinate (substance) 地考喹酯 deccox decoquinato
Molecular Weight (g/mol): 417.55
Molecular Formula: C24H35NO5
InChi Key: JHAYEQICABJSTP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC1=C(OCC)C=C2NC=C(C(=O)OCC)C(=O)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results