missing translation for 'onlineSavingsMsg'
Learn More

Decon™ LeStreak™ Microstreakers

Perfect for inoculating or transferring microbiological samples

$229.28 - $910.16

Specifications

Length (English) 10.75 in.
Length (Metric) 27.31 cm
Material Nichrome
Sterility Non-sterile
Product Type Inoculating Streaker
View More Specs

Products 5
Catalog Number Mfr. No. Volume Calibrated Diameter (Metric) Inside Price Quantity  
Catalog Number Mfr. No. Volume Calibrated Diameter (Metric) Inside Price Quantity  
22-032097
View Documents
Decon™
MP1825
3.1 μL No 0.29 cm
Pack of 5 Each for $229.28
Only null left
 
22-032098
View Documents
Decon™
MP1845
2.45 μL No 0.22 cm
Pack of 5 Each for $229.28
Only null left
 
22-032100
missing translation for 'viewDocuments'
Decon™
MP1855
1 μL Yes 0.4 cm
Pack of 5 Each for $868.47
Only null left
 
22-032096
missing translation for 'viewDocuments'
Decon™
DCMP1805
5.45 μL No 0.35 cm
Pack of 5 Each for $229.28
Only null left
 
22-032101
missing translation for 'viewDocuments'
Decon™
MP1875
10 μL Yes 0.14 cm
Pack of 5 Each for $910.16
Only null left
 
Description

Description

Perfect for inoculating or transferring microbiological samples

  • Use to inoculate or transfer microbiological samples
  • Made of high-quality twisted nichrome wire permanently attached to lightweight insulated aluminum handles
  • Slide smoothly across agar surfaces without tearing or gouging
  • Nichrome wire cools rapidly after flaming
  • 1 and 10μL loops are individually calibrated in accordance with ASM/APHA via Evans Blue Dye Test
  • Noncalibrated microstreakers are color coded for easy identification

Certifications

Calibration certificates provided for 1μL and 10μL loops

Specifications

Specifications

10.75 in.
Nichrome
Inoculating Streaker
27.31 cm
Non-sterile
Videos
SDS
Documents

Documents