missing translation for 'onlineSavingsMsg'
Learn More

Decon™ DeSCAL™ Concentrate
GREENER_CHOICE

Acidic detergent designed to remove hard-water mineral deposits

$90.80 - $4,357.00

Specifications

Product Type Concentrated Descaler
For Use With (Application) Acidic detergent designed to remove hard-water mineral deposits, effectively removes urine scale from animal cages
For Use With (Equipment) Use in automatic glassware washers, process equipment, stills, autoclaves and baths
pH 1
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
0433546
GREENER_CHOICE
View Documents Promotion Details
Decon™
DC4030
114 L
Each for $4,357.00
Only null left
 
04355103
GREENER_CHOICE
View Documents Promotion Details
Decon™
DC4004
3.8 L
Case of 4 Each for $1,215.37
Only null left
 
0435536
GREENER_CHOICE
View Documents Promotion Details
Decon™
DC4001
1 L
Each for $90.80
Case of 12 Each for $1,089.60
Only null left
 
Description

Description

  • Effectively removes urine scale from animal cages
  • Use in automatic glassware washers, process equipment, stills, autoclaves and baths
Specifications

Specifications

Concentrated Descaler
Acidic detergent designed to remove hard-water mineral deposits, effectively removes urine scale from animal cages
Use in automatic glassware washers, process equipment, stills, autoclaves and baths
1
Videos
SDS
Documents

Documents