Learn More
Decon™ DeSCAL™ Concentrate
GREENER_CHOICE
Acidic detergent designed to remove hard-water mineral deposits
Supplier:  Decon™ DC4004
 View more versions of this product
Catalog No. 04355103
Only null left
Description
Description
  • Effectively removes urine scale from animal cages
  • Use in automatic glassware washers, process equipment, stills, autoclaves and baths
Specifications
Chemical Identifiers
Concentrated Descaler
Acidic detergent designed to remove hard-water mineral deposits, effectively removes urine scale from animal cages
Use in automatic glassware washers, process equipment, stills, autoclaves and baths
1
Specifications
Concentrated Descaler
3.8 L
Use in automatic glassware washers, process equipment, stills, autoclaves and baths
1 gal.
Acidic detergent designed to remove hard-water mineral deposits, effectively removes urine scale from animal cages
1
Product Suggestions
Product Suggestions