missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decon™ Contrex™ AW Powdered Autowasher Detergent
Formulated for use in all automatic labware washing machines
$118.07 - $1174.57
Specifications
| Product Type | Detergent |
|---|---|
| Color-coded | No |
| Form | Powder |
| pH | 11.5 to 12.5 |
| Scent | Regular Scent |
Description
- Concentrated powder
- Low-foaming, completely soluble, and free-rinsing
- Biodegradable
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
- Non-toxic: Item is non-toxic
Specifications
| Detergent | |
| Powder | |
| Regular Scent | |
| White |
| No | |
| 11.5 to 12.5 | |
| Non-sterile | |
| For use in all automatic labware washing machines |