missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decanoyl Chloride 98.0+%, TCI America™
$31.68 - $201.32
Chemical Identifiers
| CAS | 112-13-0 |
|---|---|
| Molecular Formula | C10H19ClO |
| Molecular Weight (g/mol) | 190.711 |
| MDL Number | MFCD00000771 |
| InChI Key | IPIVAXLHTVNRBS-UHFFFAOYSA-N |
| Synonym | capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n |
| PubChem CID | 66982 |
| IUPAC Name | decanoyl chloride |
| SMILES | CCCCCCCCCC(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D002725ML
|
TCI America
D002725ML |
25 mL |
Each for $31.68
|
|
|||||
|
D0027100ML
|
TCI America
D0027100ML |
100 mL |
Each for $70.84
|
|
|||||
|
D0027500ML
|
TCI America
D0027500ML |
500 mL |
Each for $201.32
|
|
|||||
Chemical Identifiers
| 112-13-0 | |
| 190.711 | |
| IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
| 66982 | |
| CCCCCCCCCC(=O)Cl |
| C10H19ClO | |
| MFCD00000771 | |
| capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
| decanoyl chloride |
Specifications
| 112-13-0 | |
| 230°C | |
| MFCD00000771 | |
| 3265 | |
| IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
| decanoyl chloride | |
| 66982 | |
| ≥98.0% (GC,T) | |
| Decanoyl Chloride |
| Yellow | |
| C10H19ClO | |
| 25 mL | |
| capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
| CCCCCCCCCC(=O)Cl | |
| 190.711 | |
| 190.71 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes