missing translation for 'onlineSavingsMsg'
Learn More

Decanoyl Chloride 98.0+%, TCI America™

Supplier:  TCI America D0027100ML

 View more versions of this product

Catalog No. D0027100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Decanoyl Chloride
Yellow
C10H19ClO
100 mL
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
CCCCCCCCCC(=O)Cl
190.711
190.71
Liquid
112-13-0
230°C
MFCD00000771
3265
IPIVAXLHTVNRBS-UHFFFAOYSA-N
decanoyl chloride
66982
≥98.0% (GC,T)

Chemical Identifiers

112-13-0
190.711
IPIVAXLHTVNRBS-UHFFFAOYSA-N
66982
CCCCCCCCCC(=O)Cl
C10H19ClO
MFCD00000771
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
decanoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0632

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions