Learn More
Decanoic acid, 99%
CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Supplier: Thermo Scientific Chemicals 167270010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Decanoic acid | |
| 30.0°C to 32.0°C | |
| 268.0°C to 270.0°C | |
| 99% | |
| CH3(CH2)8CO2H | |
| 02, II, 309 | |
| capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid | |
| GHVNFZFCNZKVNT-UHFFFAOYSA-N | |
| decanoic acid | |
| 2969 | |
| 172.27 | |
| 0.8900g/mL | |
| 150°C | |
| Glass bottle | |
| 0.89 |
| 99+% | |
| 334-48-5 | |
| White | |
| C10H20O2 | |
| MFCD00004441 | |
| 15, 1760 | |
| Solubility in water: 0.15g/L (20°C). Other solubilities: soluble in dilute hno3, soluble in ethanol, ether, chloroform, benzene and, carbon disulfide | |
| CCCCCCCCCC(=O)O | |
| 172.268 | |
| CHEBI:30813 | |
| 99% | |
| 1 kg | |
| Authentic | |
| Crystalline Solid |
Chemical Identifiers
| 334-48-5 | |
| 172.268 | |
| GHVNFZFCNZKVNT-UHFFFAOYSA-N | |
| 2969 | |
| decanoic acid |
| C10H20O2 | |
| MFCD00004441 | |
| capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid | |
| CHEBI:30813 | |
| CCCCCCCCCC(=O)O |
Safety and Handling
GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 206-376-4
RTECSNumber : HD9100000
TSCA : TSCA
RUO – Research Use Only