missing translation for 'onlineSavingsMsg'
Learn More

Decanoic acid, 99%

Catalog No. p-98568
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167271000 100 g Glass bottle
AC167275000 500 g Glass bottle
AC167270010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167271000 Supplier Thermo Scientific Chemicals Supplier No. 167271000
Only null left

CAS: 334-48-5 | C10H20O2 | 172.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 334-48-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00004441
InChI Key GHVNFZFCNZKVNT-UHFFFAOYSA-N
Synonym capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
PubChem CID 2969
ChEBI CHEBI:30813
IUPAC Name decanoic acid
SMILES CCCCCCCCCC(=O)O

Specifications

Melting Point 30.0°C to 32.0°C
Boiling Point 268.0°C to 270.0°C
Color White
Assay Percent Range 99%
Linear Formula CH3(CH2)8CO2H
Beilstein 02, II, 309
Merck Index 15, 1760
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: soluble in dilute hno3, soluble in ethanol, ether, chloroform, benzene and, carbon disulfide
Formula Weight 172.27
Percent Purity 99%
Density 0.8900g/mL
Quantity 100 g
Flash Point 150°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Specific Gravity 0.89
Chemical Name or Material Decanoic acid, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-376-4
RTECSNumber HD9100000
TSCA TSCA

RUO – Research Use Only