missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decanenitrile 98.0+%, TCI America™
$55.92 - $417.56
Chemical Identifiers
| CAS | 1975-78-6 |
|---|---|
| Molecular Formula | C10H19N |
| Molecular Weight (g/mol) | 153.269 |
| MDL Number | MFCD00039505 |
| InChI Key | HBZDPWBWBJMYRY-UHFFFAOYSA-N |
| Synonym | decanonitrile, caprinitrile, 1-cyanononane, n-decanenitrile, nonyl cyanide, n-nonyl cyanide, unii-1602vq5ivc, 1-decanenitrile, nitrile 10 d, decanoic acid nitrile |
| PubChem CID | 74791 |
| IUPAC Name | decanenitrile |
| SMILES | CCCCCCCCCC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D242625ML
|
TCI America
D242625ML |
25 mL |
Each for $55.92
|
|
|||||
|
D2426100ML
|
TCI America
D2426100ML |
100 mL |
Each for $141.67
|
|
|||||
|
D2426500ML
|
TCI America
D2426500ML |
500 mL |
Each for $417.56
|
|
|||||
Chemical Identifiers
| 1975-78-6 | |
| 153.269 | |
| HBZDPWBWBJMYRY-UHFFFAOYSA-N | |
| 74791 | |
| CCCCCCCCCC#N |
| C10H19N | |
| MFCD00039505 | |
| decanonitrile, caprinitrile, 1-cyanononane, n-decanenitrile, nonyl cyanide, n-nonyl cyanide, unii-1602vq5ivc, 1-decanenitrile, nitrile 10 d, decanoic acid nitrile | |
| decanenitrile |
Specifications
| 1975-78-6 | |
| Yellow | |
| C10H19N | |
| 25 mL | |
| HBZDPWBWBJMYRY-UHFFFAOYSA-N | |
| decanenitrile | |
| 74791 | |
| ≥98.0% (GC) | |
| Decanenitrile |
| -15°C | |
| 237°C | |
| MFCD00039505 | |
| decanonitrile, caprinitrile, 1-cyanononane, n-decanenitrile, nonyl cyanide, n-nonyl cyanide, unii-1602vq5ivc, 1-decanenitrile, nitrile 10 d, decanoic acid nitrile | |
| CCCCCCCCCC#N | |
| 153.269 | |
| 153.27 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1507
TSCA : Yes