accessibility menu, dialog, popup

UserName

CAS RN 1975-78-6

H3CN

CAS RN 1975-78-6

IUPAC Name: decanenitrile
Synonyms: decanenitrile
Molecular Weight (g/mol): 153.27
Molecular Formula: C10H19N
InChi Key: HBZDPWBWBJMYRY-UHFFFAOYSA-N
SMILES: CCCCCCCCCC#N
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results