missing translation for 'onlineSavingsMsg'
Learn More

Decanamide 98.0+%, TCI America™

Catalog No. C133110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C133110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C133110G Supplier TCI America Supplier No. C133110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2319-29-1
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.28
MDL Number MFCD00025541
InChI Key TUTWLYPCGCUWQI-UHFFFAOYSA-N
Synonym Decanoylamide
PubChem CID 75347
ChEBI CHEBI:38833
IUPAC Name decanamide
SMILES CCCCCCCCCC(N)=O

Specifications

Melting Point 98°C
Color White
Quantity 10 g
Formula Weight 171.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Decanamide
EINECSNumber (2)-0824
TSCA No