accessibility menu, dialog, popup

UserName

H3CNH2O

CAS RN 2319-29-1

IUPAC Name: decanamide
Synonyms: decanamide capramide
Molecular Weight (g/mol): 171.28
Molecular Formula: C10H21NO
InChi Key: TUTWLYPCGCUWQI-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results