missing translation for 'onlineSavingsMsg'
Learn More

Decanal 97.0+%, TCI America™

Catalog No. p-7125993
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D003225ML 25 mL
D0032500ML 500 mL
2 options

Catalog No. D003225ML

Supplier: TCI America D003225ML

Only null left

Chemical Identifiers

CAS 112-31-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00007031
InChI Key KSMVZQYAVGTKIV-UHFFFAOYSA-N
Synonym decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde
PubChem CID 8175
ChEBI CHEBI:31457
IUPAC Name decanal
SMILES CCCCCCCCCC=O

Specifications

Melting Point -5°C
Color Colorless
Boiling Point 208°C
Quantity 25 mL
UN Number 3082
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Decanal
EINECSNumber (2)-0494
RTECSNumber HD6000000
TSCA Yes
Recommended Storage Refrigerator