Learn More
StatLab™ Decal Stat Decalcifier
Decalcifies the average bone marrow biopsy in less than one hour.
$50.64 - $659.89
Specifications
Solution Type Decalcifier
View More Specs
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
STL121232
View Documents Promotion Details
StatLab™
121232
4 x 32 oz. Bottle
Each for $50.64
Case of 4 Each for $202.56
Only null left
STL12125
View Documents Promotion Details
StatLab™
12125
5 gal. Cube
Each for $659.89
Only null left
Description
Description
  • Can be used on all types of calcified tissue
  • Especially effective on bone marrow samples
  • Cellular and nuclear details remain crisp and clear
Specifications
Specifications
Decalcifier