Learn More
StatLab™ Decal Stat Decalcifier
Catalog No. STL12125
Change view
Click to view available options
Quantity:
4 x 32 oz. Bottle
5 gal. Cube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL12125 5 gal. Cube
STL121232 4 x 32 oz. Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL12125 Supplier StatLab™ Supplier No. 12125
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, in contact with skin, or inhaled.
Decalcifies the average bone marrow biopsy in less than one hour.
  • Can be used on all types of calcified tissue
  • Especially effective on bone marrow samples
  • Cellular and nuclear details remain crisp and clear
Specifications
Quantity 5 gal. Cube
Solution Type Decalcifier