missing translation for 'onlineSavingsMsg'
Learn More

Décafluorobiphényl 98,0+%, TCI America™

Numéro de catalogue. P08085G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P08085G 5 g
P080825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P08085G Fournisseur TCI America Code fournisseur P08085G

Identifiants chimiques

CAS 434-90-2
Molecular Formula C12F10
Molecular Weight (g/mol) 334.116
MDL Number MFCD00000292
InChI Key ONUFSRWQCKNVSL-UHFFFAOYSA-N
Synonym decafluorobiphenyl, perfluorobiphenyl, biphenyl, decafluoro, perfluorodiphenyl, 1,1'-biphenyl, 2,2',3,3',4,4',5,5',6,6'-decafluoro, 1,2,3,4,5-pentafluoro-6-2,3,4,5,6-pentafluorophenyl benzene, decafluoro biphenyl, pubchem9114, acmc-1ctuw
PubChem CID 67949
IUPAC Name 1,2,3,4,5-pentafluoro-6-(2,3,4,5,6-pentafluorophényl)benzène
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F

Spécifications

Melting Point 70°C
Color Blanc-jaune
Boiling Point 92°C
Quantity 5 g
UN Number 3152
Formula Weight 334.12
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material Decafluorobiphenyl